Learn More
Medchemexpress LLC HY-107371 50mg Medchemexpress, 6-Bromo-2-hydroxy-3-methoxybenzaldehyde CAS:20035-41-0 Purity:>98%
SDP
Catalog No. 502080702
Encompass_Preferred
Medchemexpress, HY-107371 50mg 6-Bromo-2-hydroxy-3-methoxybenzaldehyde CAS:20035-41-0 6-Bromo-2-hydroxy-3-methoxybenzaldehyde (NSC95682) is an IRE-1α inhibitor with an IC50 of 0.08 μM, extracted from patent WO 2008154484 A1, IRE-lα inhibitor compound 3-5. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0702 Supplier Medchemexpress LLC Supplier No. HY10737150MG
Medchemexpress, HY-107371 50mg 6-Bromo-2-hydroxy-3-methoxybenzaldehyde CAS:20035-41-0 6-Bromo-2-hydroxy-3-methoxybenzaldehyde (NSC95682) is an IRE-1α inhibitor with an IC50 of 0.08 μM, extracted from patent WO 2008154484 A1, IRE-lα inhibitor compound 3-5. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.