Learn More
Medchemexpress LLC 6-bromo-2-hydroxy-3-methoxybenzaldehyde | 20035-41-0 | MFCD00046152 | 100.0% | 231.04 | C8H7BrO3 | 1 ML
SDP
Catalog No. 5000241340
Encompass_Preferred
6-Bromo-2-hydroxy-3-methoxybenzaldehyde is a small-molecule research reagent reported as an inhibitor of IRE1α for studies of the unfolded protein response. It is supplied as a high-purity solid in multiple weights or as a 10 mM solution in DMSO, and is characterized by formula C8H7BrO3 and molecular weight 231.04.
  • High purity (99.96%) suitable for biochemical and cellular assays.
  • Available as solid (multiple sizes) and as a 10 mM solution in DMSO for convenience.
  • Reported activity as an IRE1α inhibitor useful for unfolded protein response research.
  • Stable when stored at recommended temperatures for powder and solutions.
  • Molecular formula C8H7BrO3 and molecular weight 231.04 aid experimental planning.
Catalog No. 50-002-41340 Supplier Medchemexpress LLC Supplier No. HY10737110MM 1ML
6-Bromo-2-hydroxy-3-methoxybenzaldehyde is a small-molecule research reagent reported as an inhibitor of IRE1α for studies of the unfolded protein response. It is supplied as a high-purity solid in multiple weights or as a 10 mM solution in DMSO, and is characterized by formula C8H7BrO3 and molecular weight 231.04.
  • High purity (99.96%) suitable for biochemical and cellular assays.
  • Available as solid (multiple sizes) and as a 10 mM solution in DMSO for convenience.
  • Reported activity as an IRE1α inhibitor useful for unfolded protein response research.
  • Stable when stored at recommended temperatures for powder and solutions.
  • Molecular formula C8H7BrO3 and molecular weight 231.04 aid experimental planning.