Learn More
Medchemexpress LLC 6-Bromo-2-naphthol | 15231-91-1 | 223.07 | 25 G
SDP
Catalog No. 5000449952
Encompass_Preferred
6-Bromo-2-naphthol is an RTP (real-time polymerase chain reaction) probe that forms a complex with β-cyclodextrin (β-CD). It enables real-time monitoring of PCR reactions and quantification of specific nucleic acid sequences. This compound is used in gene expression analysis, SNP genotyping, and pathogen detection. It can also serve as an intermediate for synthesizing antibacterial azo dyes.
  • Forms a 1:1 or 2:1 complex with β-cyclodextrin
  • Emits phosphorescent signal at room temperature
  • Suitable for real-time monitoring of PCR reactions
  • Used for quantification of specific nucleic acid sequences
  • Inhibits oxygen quenching through hydrophobic interactions
  • Derivatives show antibacterial activity against *Staphylococcus aureus* and *Escherichia coli*
Catalog No. 50-004-49952 Supplier Medchemexpress LLC Supplier No. 25GHYW001952
6-Bromo-2-naphthol is an RTP (real-time polymerase chain reaction) probe that forms a complex with β-cyclodextrin (β-CD). It enables real-time monitoring of PCR reactions and quantification of specific nucleic acid sequences. This compound is used in gene expression analysis, SNP genotyping, and pathogen detection. It can also serve as an intermediate for synthesizing antibacterial azo dyes.
  • Forms a 1:1 or 2:1 complex with β-cyclodextrin
  • Emits phosphorescent signal at room temperature
  • Suitable for real-time monitoring of PCR reactions
  • Used for quantification of specific nucleic acid sequences
  • Inhibits oxygen quenching through hydrophobic interactions
  • Derivatives show antibacterial activity against *Staphylococcus aureus* and *Escherichia coli*