Learn More
Medchemexpress LLC 6-Bromo-2-naphthyl β-D-glucopyranoside | 15548-61-5 | 385.21 | 1 G
SDP
Catalog No. 5000350661
Encompass_Preferred
6-Bromo-2-naphthyl β-D-glucopyranoside is a biochemical reagent that can be used as a biological material or organic compound for life science related research. This product is for research use only.
  • Biochemical reagent for life science research
  • For research use only
  • Solid appearance
  • White to light yellow color
  • Store at -20°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months; -20°C for 1 month
  • Shipping at room temperature in continental US
Catalog No. 50-003-50661 Supplier Medchemexpress LLC Supplier No. HY1377781G
6-Bromo-2-naphthyl β-D-glucopyranoside is a biochemical reagent that can be used as a biological material or organic compound for life science related research. This product is for research use only.
  • Biochemical reagent for life science research
  • For research use only
  • Solid appearance
  • White to light yellow color
  • Store at -20°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months; -20°C for 1 month
  • Shipping at room temperature in continental US