Learn More
Medchemexpress LLC 6-Bromo-3,4-dihydro-1(2H)-naphthalenone | 66361-67-9 | 225.08 | 500 MG
SDP
Catalog No. 5000447414
Encompass_Preferred
6-Bromo-3,4-dihydro-1(2H)-naphthalenone is a biochemical reagent suitable for life science-related research. This product is for research use only and not for sale to patients.
  • Can be used as a biological material or organic compound
  • Soluble in DMSO up to 100 mg/mL
  • Powder can be stored at -20°C for 3 years or 4°C for 2 years
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-47414 Supplier Medchemexpress LLC Supplier No. HYW008321500MG
6-Bromo-3,4-dihydro-1(2H)-naphthalenone is a biochemical reagent suitable for life science-related research. This product is for research use only and not for sale to patients.
  • Can be used as a biological material or organic compound
  • Soluble in DMSO up to 100 mg/mL
  • Powder can be stored at -20°C for 3 years or 4°C for 2 years
  • In solvent, store at -80°C for 6 months or -20°C for 1 month