Learn More
Medchemexpress LLC 6-bromo-3-fluoropyridine-2-carbaldehyde | 885267-36-7 | MFCD07781224 | 99.9% | 204.00 g/mol | C6H3BrFNO | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW058423
Encompass_Preferred
6-Bromo-3-fluoropyridine-2-carbaldehyde is an organic intermediate used in medicinal chemistry and small-molecule synthesis. It features a brominated and fluorinated pyridine ring with an aldehyde functionality, enabling a range of derivatization and coupling reactions. CAS 885267-36-7; molecular formula C6H3BrFNO; molecular weight 204.00 g/mol.
  • Used as an intermediate in medicinal chemistry and small-molecule synthesis.
  • White to light yellow solid appearance.
  • High purity: 99.9% (GC).
  • Molecular formula C6H3BrFNO; molecular weight 204.00 g/mol.
  • Suitable for aldehyde-based derivatization and coupling reactions.
Catalog No. 50-002-08564