Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-bromo-3-fluoropyridine-2-carbaldehyde | 885267-36-7 | MFCD07781224 | 99.9% | 204.00 g/mol | C6H3BrFNO | 5 G
SDP

Catalog No. 5000209924
Encompass_Preferred

6-bromo-3-fluoropyridine-2-carbaldehyde is a substituted pyridine aldehyde used as a chemical intermediate in the synthesis of pharmaceuticals and other biologically active compounds. It is supplied as a solid and is commonly used in medicinal chemistry to construct heteroaromatic frameworks.

  • High purity: 99.9% assay.
  • Molecular formula: C6H3BrFNO; molecular weight: 204.00 g/mol.
  • Appearance: white to light yellow solid.
  • Storage: -20°C sealed, away from moisture; in solvent: -80°C for up to 6 months, -20°C for 1 month.
  • Typical application: chemical intermediate for drug and active compound synthesis.

Catalog No. 50-002-09924 Supplier Medchemexpress LLC Supplier No. HYW0584235G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-bromo-3-fluoropyridine-2-carbaldehyde is a substituted pyridine aldehyde used as a chemical intermediate in the synthesis of pharmaceuticals and other biologically active compounds. It is supplied as a solid and is commonly used in medicinal chemistry to construct heteroaromatic frameworks.

  • High purity: 99.9% assay.
  • Molecular formula: C6H3BrFNO; molecular weight: 204.00 g/mol.
  • Appearance: white to light yellow solid.
  • Storage: -20°C sealed, away from moisture; in solvent: -80°C for up to 6 months, -20°C for 1 month.
  • Typical application: chemical intermediate for drug and active compound synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.