Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-bromo-3-fluoropyridine-2-carbaldehyde | 885267-36-7 | MFCD07781224 | 99.9% | 204.00 g/mol | C6H3BrFNO | 500 MG
SDP

Catalog No. 5000210005
Encompass_Preferred

6-bromo-3-fluoropyridine-2-carbaldehyde is a pyridine-derived aldehyde used as a synthetic intermediate in medicinal chemistry research. It is supplied as a solid with high chemical purity and characterized by standard analytical testing.

  • Used as a synthetic intermediate in medicinal chemistry
  • Cas number 885267-36-7
  • Molecular formula C6H3BrFNO
  • Molecular weight 204.00 g/mol
  • Purity (GC) 99.92%
  • Appearance white to light yellow solid
  • Storage -20°C, sealed and protected from moisture; in solvent: -80°C up to 6 months, -20°C up to 1 month
  • Typical supplied quantity 500 mg

Catalog No. 50-002-10005 Supplier Medchemexpress LLC Supplier No. HYW058423500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-bromo-3-fluoropyridine-2-carbaldehyde is a pyridine-derived aldehyde used as a synthetic intermediate in medicinal chemistry research. It is supplied as a solid with high chemical purity and characterized by standard analytical testing.

  • Used as a synthetic intermediate in medicinal chemistry
  • Cas number 885267-36-7
  • Molecular formula C6H3BrFNO
  • Molecular weight 204.00 g/mol
  • Purity (GC) 99.92%
  • Appearance white to light yellow solid
  • Storage -20°C, sealed and protected from moisture; in solvent: -80°C up to 6 months, -20°C up to 1 month
  • Typical supplied quantity 500 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.