Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-bromo-3-pyridinemethanol | 122306-01-8 | MFCD04974508 | >98.0% | 188.02 g/mol | C6H6BrNO | 1 ML
SDP

Catalog No. 5000302259
Encompass_Preferred

(6-Bromopyridin-3-yl)methanol is a brominated pyridinemethanol used as a biochemical reagent and synthetic intermediate in life-science and pharmaceutical research. It is supplied in assay-ready solution formats and characterized for storage and formulation stability to support synthesis and biological testing.

  • Used as a biochemical assay reagent and synthetic intermediate.
  • Molecular formula: C6H6BrNO.
  • Molecular weight: 188.02 g/mol.
  • Typical solution formats available (for example, 10 mM in solvent) for assay use.
  • Storage recommendations: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Solubility: soluble in formulation vehicles (eg, 10% DMSO/40% PEG300/5% Tween-80/45% saline) at ≥2.5 mg/mL (~13.3 mM).

Catalog No. 50-003-02259 Supplier Medchemexpress LLC Supplier No. HYW01611610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(6-Bromopyridin-3-yl)methanol is a brominated pyridinemethanol used as a biochemical reagent and synthetic intermediate in life-science and pharmaceutical research. It is supplied in assay-ready solution formats and characterized for storage and formulation stability to support synthesis and biological testing.

  • Used as a biochemical assay reagent and synthetic intermediate.
  • Molecular formula: C6H6BrNO.
  • Molecular weight: 188.02 g/mol.
  • Typical solution formats available (for example, 10 mM in solvent) for assay use.
  • Storage recommendations: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Solubility: soluble in formulation vehicles (eg, 10% DMSO/40% PEG300/5% Tween-80/45% saline) at ≥2.5 mg/mL (~13.3 mM).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.