Learn More
Medchemexpress LLC 6-bromo-3-pyridinemethanol | 122306-01-8 | MFCD04974508 | 98.8% | 188.02 g/mol | C6H6BrNO | 10 G
SDP
Catalog No. 5000303218
Encompass_Preferred
(6-Bromopyridin-3-yl)methanol is a brominated pyridine alcohol used as an organic building block and intermediate in medicinal chemistry and biochemical research. It is employed for constructing heterocyclic scaffolds and introducing a hydroxymethyl functionality in multi-step organic syntheses.
Catalog No. 50-003-03218 Supplier Medchemexpress LLC Supplier No. HYW01611610G
(6-Bromopyridin-3-yl)methanol is a brominated pyridine alcohol used as an organic building block and intermediate in medicinal chemistry and biochemical research. It is employed for constructing heterocyclic scaffolds and introducing a hydroxymethyl functionality in multi-step organic syntheses.