Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Bromo-4-chloroquinoline | 65340-70-7 | 99.96% | C9H5BrClN | 25 G
SDP

Catalog No. 5000359063
Encompass_Preferred

6-Bromo-4-chloroquinoline is a drug intermediate used in the synthesis of various active compounds. It has a molecular weight of 242.50.

  • Appearance: Solid
  • Color: White to light yellow
  • Molecular weight: 242.50

Catalog No. 50-003-59063 Supplier Medchemexpress LLC Supplier No. 25GHY75144
May include imposed supplier surcharges.
Add to Cart
Add to Cart

6-Bromo-4-chloroquinoline is a drug intermediate used in the synthesis of various active compounds. It has a molecular weight of 242.50.

  • Appearance: Solid
  • Color: White to light yellow
  • Molecular weight: 242.50

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.