Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Bromo-4-fluoro-1-isopropyl-2-methyl-1H-benzo[d]imidazole | 1231930-33-8 | 99.4% | C11H12BrFN2 | 500MG
SDP

Catalog No. 5000434446
Encompass_Preferred

Abemaciclib Impurity 1 is a drug intermediate designed for the synthesis of various active compounds. It is supplied as an off-white to yellow solid with a purity of 99.4%.

  • A drug intermediate for chemical synthesis
  • Appearance as an off-white to yellow solid
  • Purity of 99.4%
  • Powder storage at -20°C for 3 years or 4°C for 2 years
  • In-solvent storage at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-34446 Supplier Medchemexpress LLC Supplier No. HY78066500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Abemaciclib Impurity 1 is a drug intermediate designed for the synthesis of various active compounds. It is supplied as an off-white to yellow solid with a purity of 99.4%.

  • A drug intermediate for chemical synthesis
  • Appearance as an off-white to yellow solid
  • Purity of 99.4%
  • Powder storage at -20°C for 3 years or 4°C for 2 years
  • In-solvent storage at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.