Learn More
Sigma Aldrich 6-Bromo-4-hydroxy-2-methylquinoline
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Reactant involved in synthesis of molecules for biological studies including:
- Cytotoxicity and insecticidal activities of harmine derivatives
- Isoquinolines, α-carbolines, and 3-deazapurines via oxidative decarboxylation
- Cytotoxic evaluation of 1,3-di- and 1,3,9-trisubstituted α-carbolinesimplicated in eosinophilia-myalgia syndrome associated with L-tryptophan ingestion
Specifications
Specifications
| CAS | 103030-28-0 |
| Molecular Formula | C10H8BrNO |
| Linear Formula | C10H8BrNO |
| Quantity | 100 mg |
| MDL Number | MFCD00484433 |
| Molecular Weight (g/mol) | 238.08 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.