Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo-4-hydroxycoumarin 98.0+%, TCI America™
SDP

Catalog No. B46995G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46995G 5 g
B46991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46995G Supplier TCI America Supplier No. B46995G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4139-61-1
Molecular Formula C9H5BrO3
Molecular Weight (g/mol) 241.04
MDL Number MFCD00239372
InChI Key KNMCTCABMSGXGR-UHFFFAOYSA-N
PubChem CID 54691404
IUPAC Name 6-bromo-4-hydroxychromen-2-one
SMILES C1=CC2=C(C=C1Br)C(=CC(=O)O2)O

Specifications

Melting Point 277°C
Quantity 5 g
Formula Weight 241.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-4-hydroxycoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.