Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Bromobenzo[d]thiazole | 53218-26-1 | 214.08 | 5 G
SDP

Catalog No. 5000445738
Encompass_Preferred

6-Bromobenzo[d]thiazole is a biochemical reagent suitable for life science related research. It is intended for research use only and is not sold to patients.

  • Off-white to yellow solid appearance
  • Purity of >99.0%
  • Molecular weight of 214.08
  • Powder storage at -20°C for 3 years or 4°C for 2 years
  • In solvent storage at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-45738 Supplier Medchemexpress LLC Supplier No. HYW0031825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Bromobenzo[d]thiazole is a biochemical reagent suitable for life science related research. It is intended for research use only and is not sold to patients.

  • Off-white to yellow solid appearance
  • Purity of >99.0%
  • Molecular weight of 214.08
  • Powder storage at -20°C for 3 years or 4°C for 2 years
  • In solvent storage at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.