Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromobenzothiazole, 97%

Catalog No. AAH5282303
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5282303 1 g
AAH52823MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5282303 Supplier Thermo Scientific Chemicals Supplier No. H5282303
Only null left
Add to Cart

CAS: 53218-26-1 | C7H4BrNS | 214.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53218-26-1
Molecular Formula C7H4BrNS
Molecular Weight (g/mol) 214.08
MDL Number MFCD04115372
InChI Key YJOUISWKEOXIMC-UHFFFAOYSA-N
Synonym 6-bromobenzothiazole, 6-bromobenzo d thiazole, benzothiazole, 6-bromo, 6-bromo-benzothiazole, 6-bromo-benzo d thiazole, pubchem21861, acmc-209l4t, 6-bromanyl-1,3-benzothiazole, benzothiazole, 6-bromo-9ci
PubChem CID 2795171
IUPAC Name 6-bromo-1,3-benzothiazole
SMILES C1=CC2=C(C=C1Br)SC=N2

Specifications

Melting Point 52°C to 54°C
Quantity 1 g
Formula Weight 214.08
Percent Purity 97%
Chemical Name or Material 6-Bromobenzothiazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.