Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromochromone-3-carbonitrile 98.0+%, TCI America™
SDP

Catalog No. B49681G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49681G 1 g
B49685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49681G Supplier TCI America Supplier No. B49681G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52817-13-7
Molecular Formula C10H4BrNO2
Molecular Weight (g/mol) 250.051
MDL Number MFCD00191842
InChI Key MGVVCEKLWMZFLS-UHFFFAOYSA-N
Synonym 6-Bromo-3-cyanochromone, 6-Bromo-4-oxo-4H-1-benzopyran-3-carbonitrile
PubChem CID 703036
IUPAC Name 6-bromo-4-oxochromene-3-carbonitrile
SMILES C1=CC2=C(C=C1Br)C(=O)C(=CO2)C#N

Specifications

Melting Point 225°C
Color Yellow
Quantity 1 g
UN Number 3439
Formula Weight 250.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromochromone-3-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.