Learn More
Medchemexpress LLC 6-Bromocoumarin | 19063-55-9 | 225.04 | 1 G
SDP
Catalog No. 5000445500
Encompass_Preferred
6-Bromocoumarin is a potential anti-bacterial agent that exhibits affinity for α and β estrogen receptors. It does not inhibit 17β-HSD1 and is intended for research use only.
  • Potential anti-bacterial agent
  • Exhibits affinity for α and β estrogen receptors
  • Does not inhibit 17β-HSD1
  • High purity of 98.51%
  • White to off-white solid appearance
  • Molecular weight of 225.04
Catalog No. 50-004-45500 Supplier Medchemexpress LLC Supplier No. HYW0051861G
6-Bromocoumarin is a potential anti-bacterial agent that exhibits affinity for α and β estrogen receptors. It does not inhibit 17β-HSD1 and is intended for research use only.
  • Potential anti-bacterial agent
  • Exhibits affinity for α and β estrogen receptors
  • Does not inhibit 17β-HSD1
  • High purity of 98.51%
  • White to off-white solid appearance
  • Molecular weight of 225.04