Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromoimidazo[1,2-a]pyrimidine 98.0+%, TCI America™
SDP

Catalog No. B55991G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55991G 1 g
B55995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55991G Supplier TCI America Supplier No. B55991G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 865156-68-9
Molecular Formula C6H4BrN3
Molecular Weight (g/mol) 198.023
MDL Number MFCD09261434
InChI Key BQMWMOQCMFLRQQ-UHFFFAOYSA-N
Synonym 6-bromoimidazo 1,2-a pyrimidine, 6-bromo-imidazo 1,2-a pyrimidine, imidazo 1,2-a pyrimidine, 6-bromo, 6-bromo-4-hydroimidazo 1,2-a pyrimidine, pubchem20931, acmc-209qa4, 6-bromo-imidazo 1,2-a pyr..., 6-bromanylimidazo 1,2-a pyrimidine, imidazo 1,2-a pyrimidine,6-bromo
PubChem CID 26967622
IUPAC Name 6-bromoimidazo[1,2-a]pyrimidine
SMILES C1=CN2C=C(C=NC2=N1)Br

Specifications

Melting Point 218°C
Color White-Yellow
Quantity 1 g
Formula Weight 198.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromoimidazo[1,2-a]pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.