Learn More
Accela Chembio Inc 6-bromoisoquinoline | 25g | 34784-05-9 | MFCD04973299 | 97+% | Shelf Life: 1440 Days | Light Sensitive/nitrogen Or Argon/+4
Supplier:  Accela Chembio Inc SY00458025G
Encompass_Preferred
6-bromoisoquinoline | 25g | 34784-05-9 | MFCD04973299 | 97+% | Shelf Life: 1440 Days | Light Sensitive/nitrogen Or Argon/+4
Catalog No. 50-250-516