Learn More
Medchemexpress LLC 6-Bromoisoquinoline | 34784-05-9 | 98.2% | C9H6BrN | 25 G
SDP
Catalog No. 5000358971
Encompass_Preferred
6-Bromoisoquinoline is a biochemical reagent used as a biological material or organic compound for life science research. It serves as a synthetic intermediate for pharmaceutical synthesis.
  • Biochemical reagent for life science research
  • Synthetic intermediate for pharmaceutical synthesis
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-58971 Supplier Medchemexpress LLC Supplier No. 25GHY69092
6-Bromoisoquinoline is a biochemical reagent used as a biological material or organic compound for life science research. It serves as a synthetic intermediate for pharmaceutical synthesis.
  • Biochemical reagent for life science research
  • Synthetic intermediate for pharmaceutical synthesis
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month