Learn More
Medchemexpress LLC 6-Bromonicotinic acid | 6311-35-9 | MFCD01927100 | 202.01 | 10 G
SDP
Catalog No. 5000448604
Encompass_Preferred
6-Bromonicotinic acid is a brominated derivative of Niacin and can be used as a drug intermediate for the synthesis of Anabaseine derivatives. It is intended for research use only.
  • Brominated derivative of Niacin
  • Used as a drug intermediate
  • Synthesizes Anabaseine derivatives
Catalog No. 50-004-48604 Supplier Medchemexpress LLC Supplier No. HYW00286410G
Add to Cart
Add to Cart
6-Bromonicotinic acid is a brominated derivative of Niacin and can be used as a drug intermediate for the synthesis of Anabaseine derivatives. It is intended for research use only.
  • Brominated derivative of Niacin
  • Used as a drug intermediate
  • Synthesizes Anabaseine derivatives