Learn More
Medchemexpress LLC 6-bromopyridin-2-amine | 19798-81-3 | MFCD00137843 | 100.0% | 173.01 g·mol⁻¹ | C5H5BrN2 | 100g
SDP
Catalog No. 5000267874
Encompass_Preferred
2-Amino-6-bromopyridine is an intermediate 2-Amino-6-bromopyridine is also a PqsR ligand with a Kd value of 6 8 M in SPR assay 2-Amino-6-bromopyridine shows weak antagonistic activity 2-Amino-6-bromopyridine can be used in the synthesis of JAK2 inhibitors MSK1 inhibitors and in the research of Pseudomonas aeruginosa infection[1][2][3][4]
Catalog No. 50-002-67874 Supplier Medchemexpress LLC Supplier No. HYY1049100G
2-Amino-6-bromopyridine is an intermediate 2-Amino-6-bromopyridine is also a PqsR ligand with a Kd value of 6 8 M in SPR assay 2-Amino-6-bromopyridine shows weak antagonistic activity 2-Amino-6-bromopyridine can be used in the synthesis of JAK2 inhibitors MSK1 inhibitors and in the research of Pseudomonas aeruginosa infection[1][2][3][4]