Learn More
Medchemexpress LLC 6-bromopyridin-3-amine | 13534-97-9 | MFCD01646116 | 99.1% | 173.01 g·mol⁻¹ | C5H5BrN2 | 50 G
SDP
Catalog No. 5000300428
Encompass_Preferred
6-Bromopyridin-3-amine is an organic building block and biochemical reagent used as an intermediate in medicinal chemistry and small-molecule synthesis. It is employed for heterocycle functionalization and derivatization in research applications, offering a brominated aminopyridine scaffold for downstream transformations.
  • High purity suitable for research use (≈99.1%).
  • Molecular formula C5H5BrN2; molecular weight 173.01 g·mol⁻¹.
  • Solid form, light brown to yellow, stable at room temperature for handling and shipping.
  • Available in multiple pack sizes including 50 g for scale-up needs.
  • Commonly used as a synthetic intermediate in medicinal chemistry and organic synthesis.
Catalog No. 50-003-00428 Supplier Medchemexpress LLC Supplier No. HYW01684950G
6-Bromopyridin-3-amine is an organic building block and biochemical reagent used as an intermediate in medicinal chemistry and small-molecule synthesis. It is employed for heterocycle functionalization and derivatization in research applications, offering a brominated aminopyridine scaffold for downstream transformations.
  • High purity suitable for research use (≈99.1%).
  • Molecular formula C5H5BrN2; molecular weight 173.01 g·mol⁻¹.
  • Solid form, light brown to yellow, stable at room temperature for handling and shipping.
  • Available in multiple pack sizes including 50 g for scale-up needs.
  • Commonly used as a synthetic intermediate in medicinal chemistry and organic synthesis.