Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Bromoquinoline | 5332-25-2 | 208.06 | 25 G
SDP

Catalog No. 5000449297
Encompass_Preferred

6-Bromoquinoline is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It serves as a fine chemical and pharmaceutical intermediate, specifically as a coupling reagent.

  • Used as a biochemical reagent
  • Functions as a biological material or organic compound for life science research
  • Serves as a fine chemical and pharmaceutical intermediate
  • Acts as a coupling reagent
  • Purity: 99.0%
  • Molecular weight: 208.06
  • Appearance: <19°C Solid, >19°C Liquid
  • Color: Light yellow to brown

Catalog No. 50-004-49297 Supplier Medchemexpress LLC Supplier No. 25GHYW007403
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Bromoquinoline is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It serves as a fine chemical and pharmaceutical intermediate, specifically as a coupling reagent.

  • Used as a biochemical reagent
  • Functions as a biological material or organic compound for life science research
  • Serves as a fine chemical and pharmaceutical intermediate
  • Acts as a coupling reagent
  • Purity: 99.0%
  • Molecular weight: 208.06
  • Appearance: <19°C Solid, >19°C Liquid
  • Color: Light yellow to brown

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.