Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromoveratraldehyde, 97%

Catalog No. AAA1359106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1359106 5 g
AAA1359114 25 g
AAA1359122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1359106 Supplier Thermo Scientific Chemicals Supplier No. A1359106
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 5392-10-9 | C9H9BrO3 | 245.07 g/mol

6-Bromoveratraldehyde was used in the preparation of benzylchromenylamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromoveratraldehyde was used in the preparation of benzylchromenylamines.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5392-10-9
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.07
MDL Number MFCD00003301
InChI Key UQQROBHFUDBOOK-UHFFFAOYSA-N

Specifications

Melting Point 147°C to 152°C
Quantity 5 g
Beilstein 2052355
Sensitivity Air Sensitive
Synonym 2-methoxy-1-naphthaldehyde, 1-naphthalenecarboxaldehyde, 2-methoxy, 2-methoxy-1-naphthalenecarboxaldehyde, unii-7q7tx7tb9e, 7q7tx7tb9e, 2-methoxy-1-naphthylaldehyde, 2-methoxy-naphthalene-1-carbaldehyde, 2-methoxynaphthalenecarbaldehyde, 2-methoxy-1-naphthaldehyd, 2-methoxynaphthaldehyde
Solubility Information Insoluble in water.
SMILES COC1=CC(Br)=C(C=O)C=C1OC
IUPAC Name 2-methoxynaphthalene-1-carbaldehyde
PubChem CID 79352
Formula Weight 245.08
Percent Purity 97%
Chemical Name or Material 6-Bromoveratraldehyde
Show More Show Less
EINECSNumber 226-390-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.