Learn More
Medchemexpress LLC 6-Carboxy-X-rhodamine | 194785-18-7 | 98.43% | 50 MG
SDP
Catalog No. 5000409782
Encompass_Preferred
6-ROX is a selective fluorescent probe and potential inhibitor of COX-2. It binds to the active site of COX-2 and inhibits its conversion of arachidonic acid into prostaglandins. This compound is frequently utilized in optical imaging applications related to tumors and inflammation, thereby assisting in the detection of diseased tissues that exhibit high expression levels of COX-2.
  • Selective fluorescent probe and potential inhibitor of COX-2.
  • Binds to the active site of COX-2.
  • Inhibits conversion of arachidonic acid into prostaglandins.
  • Used in optical imaging related to tumors and inflammation.
  • Helps detect diseased tissues with high expression of COX-2.
Catalog No. 50-004-09782 Supplier Medchemexpress LLC Supplier No. HYD005350MG
6-ROX is a selective fluorescent probe and potential inhibitor of COX-2. It binds to the active site of COX-2 and inhibits its conversion of arachidonic acid into prostaglandins. This compound is frequently utilized in optical imaging applications related to tumors and inflammation, thereby assisting in the detection of diseased tissues that exhibit high expression levels of COX-2.
  • Selective fluorescent probe and potential inhibitor of COX-2.
  • Binds to the active site of COX-2.
  • Inhibits conversion of arachidonic acid into prostaglandins.
  • Used in optical imaging related to tumors and inflammation.
  • Helps detect diseased tissues with high expression of COX-2.