Learn More
Medchemexpress LLC 6-carboxy-X-rhodamine | 194785-18-7 | MFCD08705674 | 99.0% | 534.60 g/mol | C33H30N2O5 | 10 MG
SDP
Catalog No. 5000297835
Encompass_Preferred
6-Carboxy-X-rhodamine (6-ROX) is a red fluorescent dye and reported COX-2 inhibitor supplied as a high-purity research reagent. It is used for oligonucleotide labeling, fluorescence imaging, and as a reference dye in molecular assays. The compound is provided as a solid, is soluble in DMSO, and should be stored protected from light.
  • Red-emitting fluorescent probe suitable for imaging and labeling.
  • Reported COX-2 inhibitor that binds the enzyme active site.
  • High purity (99%) solid for research applications.
  • Soluble in DMSO; recommended stock preparations in DMSO.
  • Store protected from light at 4°C; in solvent store at -80°C for long-term stability.
Catalog No. 50-002-97835 Supplier Medchemexpress LLC Supplier No. HYD005310MG
6-Carboxy-X-rhodamine (6-ROX) is a red fluorescent dye and reported COX-2 inhibitor supplied as a high-purity research reagent. It is used for oligonucleotide labeling, fluorescence imaging, and as a reference dye in molecular assays. The compound is provided as a solid, is soluble in DMSO, and should be stored protected from light.
  • Red-emitting fluorescent probe suitable for imaging and labeling.
  • Reported COX-2 inhibitor that binds the enzyme active site.
  • High purity (99%) solid for research applications.
  • Soluble in DMSO; recommended stock preparations in DMSO.
  • Store protected from light at 4°C; in solvent store at -80°C for long-term stability.