Learn More
Medchemexpress LLC 6-Carboxy-X-rhodamine, succinimidyl ester | 216699-36-4 | 80.0% | 631.67 | 5 MG
SDP
Catalog No. 5000431341
Encompass_Preferred
6-Carboxy-X-rhodamine, succinimidyl ester (6-ROX, SE) is a fluorescent dye utilized for oligonucleotide labeling and automated DNA sequencing.
Catalog No. 50-004-31341 Supplier Medchemexpress LLC Supplier No. HYD10865MG
6-Carboxy-X-rhodamine, succinimidyl ester (6-ROX, SE) is a fluorescent dye utilized for oligonucleotide labeling and automated DNA sequencing.