Learn More
Cayman Chemical 6-Carboxyfluorescein, Purity: ≥98%, CAS: 3301-79-9, 1 g
SDP
Catalog No. 502985142
Encompass_Preferred
A single isomer derivative of 5(6)-carboxyfluorescein that can be used to fluorescently label biomolecules; excitation/emission maxima of 492 and 518 nm, respectively
Catalog No. 50-298-5142 Supplier Cayman Chemical Supplier No. 212331 G
A single isomer derivative of 5(6)-carboxyfluorescein that can be used to fluorescently label biomolecules; excitation/emission maxima of 492 and 518 nm, respectively