Learn More
Cayman Chemical 6-Carboxyfluorescein, Purity: ≥98%, CAS: 3301-79-9, 250 mg
SDP
Catalog No. 502985330
Encompass_Preferred
A single isomer derivative of 5(6)-carboxyfluorescein that can be used to fluorescently label biomolecules; excitation/emission maxima of 492 and 518 nm, respectively
Catalog No. 50-298-5330 Supplier Cayman Chemical Supplier No. 21233250 MG
A single isomer derivative of 5(6)-carboxyfluorescein that can be used to fluorescently label biomolecules; excitation/emission maxima of 492 and 518 nm, respectively