Learn More
Cayman Chemical 6-CarboxyfluoresceIn 500mg
SDP
Catalog No. 502985342
Encompass_Preferred
A single isomer derivative of 5(6)-carboxyfluorescein that can be used to fluorescently label biomolecules; excitation/emission maxima of 492 and 518 nm, respectively
Catalog No. 50-298-5342 Supplier Cayman Chemical Supplier No. 21233500 MG
Add to Cart
Add to Cart
A single isomer derivative of 5(6)-carboxyfluorescein that can be used to fluorescently label biomolecules; excitation/emission maxima of 492 and 518 nm, respectively