Learn More
Biotium 6-CARBOXYRhodamine 6G, HYDROCHLORIDE, SI
Catalog No. 501964801
Encompass_Preferred
6-CR6G (full name: 6-Carboxyrhodamine 6G, hydrochloride, single isomer) is the free acid form of 6-carboxyrhodamine 6G single isomer. Properties: Ex/Em (MeOH) = 520/546 nm; Orange red solid soluble in water (pH 7), DMF or DMSO; Store at 4°C and protect from light; C 27 H 27 ClN 2 O 5; MW: 494.97
Catalog No. 50-196-4801 Supplier Biotium Supplier No. 90011
Add to Cart
Edge
Add to Cart
6-CR6G (full name: 6-Carboxyrhodamine 6G, hydrochloride, single isomer) is the free acid form of 6-carboxyrhodamine 6G single isomer. Properties: Ex/Em (MeOH) = 520/546 nm; Orange red solid soluble in water (pH 7), DMF or DMSO; Store at 4°C and protect from light; C 27 H 27 ClN 2 O 5; MW: 494.97