Learn More
AdipoGen 6-CFDA
SDP
Supplier:  AdipoGen CDXC0040M100
Encompass_Preferred
Chemical. CAS 3348-03-6. Formula C25H16O9. MW 460.39. Synthetic. Used to differentiate viable cells from apoptotic cells. Live cells that are not apoptotic accumulate 6-carboxyfluorescein but do not accumulate annexin. CFDA enter cells and are hydrolysed more slowly than FDA. CFDA and FDA hydrolyses obey Michaelis-Menten kinetics with Km values of around 19 and 2 microM, respectively.
Catalog No. 50-256-1524