Learn More
AdipoGen 6-CFDA N-succinimidyl ester
SDP
Supplier:  AdipoGen CDXC0095M005
Encompass_Preferred
Chemical. CAS 150206-15-8. Formula C29H19NO11. MW 557.46. Synthetic. 6-CFDA is membrane-permeant and thus can be loaded into cells via incubation. Once inside the cells, 6-CFDA is hydrolyzed by intracellular esterases to 6-carboxyfluorescein. Reagent for continuously determining the intracellular pH in bacteria by fluorometry. Labeling human intervertebral disk cells in vitro for fluorescence microscopy.
Catalog No. 50-256-1476