Learn More
Arcticwhite LLC 6 Column Partitioned Reservoir, Pyramid Bottom, Polypropylene (PP), 47 ML, 25 Reagent Reservoirs per Case
SDP
Catalog No. NC0334261
Encompass
6 Column Partitioned Reservoir, Pyramid Bottom, Polypropylene (PP), 47 ML/Column, 25 Reagent Reservoirs per Case. The 6-column partitioned reservoir with a pyramid bottom holds samples or reagents for preparation, storage, and distribution in laboratory workflows. Format - 6-column partitioned reservoir. Bottom - Pyramid. Capacity - 47 mL/Column. Material - Polypropylene (PP). Supplied by ArcticWhite as 25 reagent reservoirs per case.
Catalog No. NC0334261 Supplier Arcticwhite LLC Supplier No. AWLSS30030
6 Column Partitioned Reservoir, Pyramid Bottom, Polypropylene (PP), 47 ML/Column, 25 Reagent Reservoirs per Case. The 6-column partitioned reservoir with a pyramid bottom holds samples or reagents for preparation, storage, and distribution in laboratory workflows. Format - 6-column partitioned reservoir. Bottom - Pyramid. Capacity - 47 mL/Column. Material - Polypropylene (PP). Supplied by ArcticWhite as 25 reagent reservoirs per case.