Learn More
Medchemexpress LLC 6-Chloro-1-hexanol | 2009-83-8 | 99.5% | 136.62 | 50 G
SDP
Catalog No. 5000404907
Encompass_Preferred
6-Chloro-1-hexanol is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.
  • Used in the preparation of its corresponding bromide by reaction with sodium bromide using N,N-dimethylformamide and dibromomethane (2:1) as a solvent.
  • Utilized in the selective elimination of 6-chloro-1-hexene from esters of 6-chloro-1-hexanol.
Catalog No. 50-004-04907 Supplier Medchemexpress LLC Supplier No. HY4168150G
6-Chloro-1-hexanol is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.
  • Used in the preparation of its corresponding bromide by reaction with sodium bromide using N,N-dimethylformamide and dibromomethane (2:1) as a solvent.
  • Utilized in the selective elimination of 6-chloro-1-hexene from esters of 6-chloro-1-hexanol.