Learn More
Medchemexpress LLC 6-Chloro-1-hexanol | 2009-83-8 | 99.50% | 136.62 | 1 ML
SDP
Catalog No. 5000404626
Encompass_Preferred
6-Chloro-1-hexanol is a biochemical reagent suitable for life science research. It can be used as a biological material or organic compound. It is for research use only and not sold to patients.
  • Used in the preparation of its corresponding bromide by reaction with sodium bromide.
  • Utilized in the selective elimination of 6-chloro-1-hexene from esters.
  • Soluble in DMSO (100 mg/mL), requiring ultrasonic.
  • Clear solution with various in vivo dissolution methods.
Catalog No. 50-004-04626 Supplier Medchemexpress LLC Supplier No. HY4168110MM/1ML
6-Chloro-1-hexanol is a biochemical reagent suitable for life science research. It can be used as a biological material or organic compound. It is for research use only and not sold to patients.
  • Used in the preparation of its corresponding bromide by reaction with sodium bromide.
  • Utilized in the selective elimination of 6-chloro-1-hexene from esters.
  • Soluble in DMSO (100 mg/mL), requiring ultrasonic.
  • Clear solution with various in vivo dissolution methods.