Learn More
Medchemexpress LLC 6-chloro-1-hexanol-d6 | 1219794-83-8 | 142.66 | C6H7D6ClO | 10 MG
SDP
Catalog No. 5000299062
Encompass_Preferred
6-Chloro-1-hexanol-d6 is the deuterium-labeled isotopologue of 6-chloro-1-hexanol used as a tracer and internal standard for quantitative analyses by NMR, GC-MS, and LC-MS. It provides a stable isotopic reference for accurate quantitation, recovery assessment, and method validation in analytical and synthetic workflows.
  • Deuterium-labeled analog of 6-chloro-1-hexanol.
  • Suitable as tracer and internal standard for NMR, GC-MS, and LC-MS.
  • Molecular formula C6H7D6ClO and molecular weight 142.66.
  • Available in small analytical pack sizes for method development.
  • Facilitates accurate quantitation and method validation.
Catalog No. 50-002-99062 Supplier Medchemexpress LLC Supplier No. HY41681S10MG
6-Chloro-1-hexanol-d6 is the deuterium-labeled isotopologue of 6-chloro-1-hexanol used as a tracer and internal standard for quantitative analyses by NMR, GC-MS, and LC-MS. It provides a stable isotopic reference for accurate quantitation, recovery assessment, and method validation in analytical and synthetic workflows.
  • Deuterium-labeled analog of 6-chloro-1-hexanol.
  • Suitable as tracer and internal standard for NMR, GC-MS, and LC-MS.
  • Molecular formula C6H7D6ClO and molecular weight 142.66.
  • Available in small analytical pack sizes for method development.
  • Facilitates accurate quantitation and method validation.