Learn More
Medchemexpress LLC 6-Chloro-1-hexanol-d6 | 1219794-83-8 | 98.0% | 142.66 | 5 MG
SDP
Catalog No. 5000404614
Encompass_Preferred
6-Chloro-1-hexanol-d6 is a deuterium-labeled analog of 6-Chloro-1-hexanol. Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs, making these compounds useful as tracers and internal standards for quantitative analysis in drug development.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-04614 Supplier Medchemexpress LLC Supplier No. HY41681S5MG
6-Chloro-1-hexanol-d6 is a deuterium-labeled analog of 6-Chloro-1-hexanol. Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs, making these compounds useful as tracers and internal standards for quantitative analysis in drug development.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.