Learn More
Medchemexpress LLC 6-Chloro-1-tetralone | 26673-31-4 | 180.63 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00431110G
Encompass_Preferred
6-Chloro-1-tetralone is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It is also an important intermediate in the synthesis of pharmaceutical compounds.
  • Can be used as a biological material or organic compound for life science-related research
  • Important intermediate in the synthesis of pharmaceutical compounds
Catalog No. 50-004-45929