Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Chloro-1-tetralone | 26673-31-4 | 180.63 | 5 G
SDP

Catalog No. 5000446303
Encompass_Preferred

6-Chloro-1-tetralone is a biochemical reagent that serves as a biological material or organic compound for life science research. It is also a key intermediate in the synthesis of pharmaceutical compounds, making it valuable for various scientific applications.

  • For research use only
  • Appearance: Liquid (Colorless to light yellow)
  • Purity: 98.25%
  • Molecular weight: 180.63
  • Storage: -20°C for 3 years (pure form); -80°C for 6 months (in solvent)

Catalog No. 50-004-46303 Supplier Medchemexpress LLC Supplier No. HYW0043115G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Chloro-1-tetralone is a biochemical reagent that serves as a biological material or organic compound for life science research. It is also a key intermediate in the synthesis of pharmaceutical compounds, making it valuable for various scientific applications.

  • For research use only
  • Appearance: Liquid (Colorless to light yellow)
  • Purity: 98.25%
  • Molecular weight: 180.63
  • Storage: -20°C for 3 years (pure form); -80°C for 6 months (in solvent)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.