Learn More
Medchemexpress LLC 6-chloro-1H-indol-3-yl-β-D-galactopyranoside | 138182-21-5 | MFCD00467206 | 99.7% | 329.73 g/mol | C14H16ClNO6 | 100 MG
SDP
Catalog No. 5000282587
Encompass_Preferred
Rose-β-D-Gal is a chromogenic β-galactosidase substrate supplied as a solid reagent for research use. Upon enzymatic cleavage it produces a pink/magenta product that enables visual detection of β-galactosidase activity in histochemical staining and enzymatic assays. Suitable for qualitative laboratory assays.
  • Chromogenic β-galactosidase substrate for visual detection.
  • Produces a pink/magenta color upon enzymatic cleavage.
  • Suitable for histochemical staining and enzymatic assays.
  • Provided as a solid reagent for convenient storage and handling.
  • High purity supports consistent assay results.
Catalog No. 50-002-82587 Supplier Medchemexpress LLC Supplier No. HYW127769100MG
Rose-β-D-Gal is a chromogenic β-galactosidase substrate supplied as a solid reagent for research use. Upon enzymatic cleavage it produces a pink/magenta product that enables visual detection of β-galactosidase activity in histochemical staining and enzymatic assays. Suitable for qualitative laboratory assays.
  • Chromogenic β-galactosidase substrate for visual detection.
  • Produces a pink/magenta color upon enzymatic cleavage.
  • Suitable for histochemical staining and enzymatic assays.
  • Provided as a solid reagent for convenient storage and handling.
  • High purity supports consistent assay results.