Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloro-2,3,4,9-tetrahydro-1H-carbazole-1-carboxamide 98.0+%, TCI America™
SDP

Catalog No. C273925MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C273925MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C273925MG Supplier TCI America Supplier No. C273925MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 49843-98-3
Molecular Formula C13H13ClN2O
Molecular Weight (g/mol) 248.71
MDL Number MFCD03009471
InChI Key FUZYTVDVLBBXDL-UHFFFAOYSA-N
Synonym selisistat, sirt1 inhibitor iii, 1h-carbazole-1-carboxamide, 6-chloro-2,3,4,9-tetrahydro, 6-chloro-2,3,4,9-tetrahydro-1h-carbazole-1-carboxamide, racemic, selisistat inn, 6-chloro-1,2,3,4,9-pentahydro-4ah-carbazolecarboxamide, ex-527 selisistat, ex 527 selisistat, d0e3lp, cambridge id 6859180
PubChem CID 5113032
IUPAC Name 6-chloro-2,3,4,9-tetrahydro-1H-carbazole-1-carboxamide
SMILES C1CC(C2=C(C1)C3=C(N2)C=CC(=C3)Cl)C(=O)N

Specifications

Melting Point 181°C
Color White
Quantity 25 mg
Formula Weight 248.71
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-2,3,4,9-tetrahydro-1H-carbazole-1-carboxamide
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.