Learn More
Medchemexpress LLC 6-Chloro-2-fluoro-9H-purine | 1651-29-2 | 172.55 | 5 G
SDP
Catalog No. 5000449041
Encompass_Preferred
6-Chloro-2-fluoro-9H-purine is a biochemical reagent used as a biological material or organic compound for life science related research. This product is for research use only and not sold to patients.
  • Used as a biochemical reagent
  • Suitable for life science research applications
  • Appears as a solid, light yellow to yellow in color
  • High purity of 99.22%
  • Ships at room temperature in continental US
  • Recommended storage at -20°C in a sealed container, away from moisture
  • When in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month in sealed containers, away from moisture
Catalog No. 50-004-49041 Supplier Medchemexpress LLC Supplier No. HYW0029245G
6-Chloro-2-fluoro-9H-purine is a biochemical reagent used as a biological material or organic compound for life science related research. This product is for research use only and not sold to patients.
  • Used as a biochemical reagent
  • Suitable for life science research applications
  • Appears as a solid, light yellow to yellow in color
  • High purity of 99.22%
  • Ships at room temperature in continental US
  • Recommended storage at -20°C in a sealed container, away from moisture
  • When in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month in sealed containers, away from moisture