Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloro-3-pyridinol, 97%, Thermo Scientific™

Catalog No. AC379440050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC379440050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC379440050 Supplier Thermo Scientific Chemicals Supplier No. 379440050

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 41288-96-4
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.54
MDL Number MFCD00234050
InChI Key KVCOOWROABTXDJ-UHFFFAOYSA-N
Synonym 2-chloro-5-hydroxypyridine, 6-chloro-3-pyridinol, 3-pyridinol, 6-chloro, 2-chloro-5-hydroxy pyridine, 2-chloro-5-pyridinol, 2-chloropyridin-5-ol, pubchem2410, zlchem 1336, 6-chloropyridine-3-ol, 6-chloro-pyridin-3-ol
PubChem CID 819821
IUPAC Name 6-chloropyridin-3-ol
SMILES OC1=CC=C(Cl)N=C1

Specifications

Melting Point 159.0°C to 161.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 129.54
Percent Purity 97%
Chemical Name or Material 6-Chloro-3-pyridinol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.