Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6''-chloro-4'-(3-chloro-2-fluorophenyl) dispiroindole-5'-carboxylic acid (3'R,4'S,5'R) | 1410737-09-5 | 98.0% | C23H21Cl2FN2O3 | 10MG
SDP

Catalog No. 5000260217
Encompass_Preferred

MDM2-IN-1 (Compound 30) is a synthetic small-molecule inhibitor of the MDM2-p53 protein-protein interaction supplied for research use in oncology and related cell biology studies. It contains the trans configuration and is intended for in vitro and preclinical investigations, including osteosarcoma research.

  • contains the trans configuration
  • inhibits MDM2-p53 interaction in biochemical and cellular assays
  • available as solid material and as a 10 mM solution in DMSO
  • high purity (HPLC): 98.0%
  • molecular formula C23H21Cl2FN2O3; molecular weight 463.33
  • supplied with certificate of analysis and safety data sheet

Catalog No. 50-002-60217 Supplier Medchemexpress LLC Supplier No. HY13068410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MDM2-IN-1 (Compound 30) is a synthetic small-molecule inhibitor of the MDM2-p53 protein-protein interaction supplied for research use in oncology and related cell biology studies. It contains the trans configuration and is intended for in vitro and preclinical investigations, including osteosarcoma research.

  • contains the trans configuration
  • inhibits MDM2-p53 interaction in biochemical and cellular assays
  • available as solid material and as a 10 mM solution in DMSO
  • high purity (HPLC): 98.0%
  • molecular formula C23H21Cl2FN2O3; molecular weight 463.33
  • supplied with certificate of analysis and safety data sheet

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.