Learn More
Medchemexpress LLC 6-chloro-4-N-cyclopropyl-4-N-(thiophen-2-ylmethyl)pyrimidine-2,4-diamine | 1252362-53-0 | MFCD23148480 | 99.8% | 280.78 g/mol | C12H13ClN4S | 100 MG
SDP
Catalog No. 5000214913
Encompass_Preferred
LRE1 is a small-molecule, specific, allosteric inhibitor of soluble adenylyl cyclase (sAC) used in biochemical and cell-based research to probe cAMP-mediated signaling pathways. It is provided with defined chemical identifiers and high purity suitable for experimental applications.
  • Specific allosteric inhibitor of soluble adenylyl cyclase
  • High purity appropriate for research use
  • Characterized molecular weight and chemical formula
  • Available in milligram-scale quantities for laboratory studies
  • Applicable to biochemical and cell-based assays probing cAMP signaling
Catalog No. 50-002-14913 Supplier Medchemexpress LLC Supplier No. HY100524100MG
LRE1 is a small-molecule, specific, allosteric inhibitor of soluble adenylyl cyclase (sAC) used in biochemical and cell-based research to probe cAMP-mediated signaling pathways. It is provided with defined chemical identifiers and high purity suitable for experimental applications.
  • Specific allosteric inhibitor of soluble adenylyl cyclase
  • High purity appropriate for research use
  • Characterized molecular weight and chemical formula
  • Available in milligram-scale quantities for laboratory studies
  • Applicable to biochemical and cell-based assays probing cAMP signaling