Learn More
Description
- Iodomethyl methyl ether (iodomethoxymethane) is a halo ether
- Product contains copper as stabilizer
- Rotational, centrifugal distortion constants and the nuclear electric quadrupole coupling constant tensor for iodomethyl methyl ether (ch3och2i) and its two deuterated species have been measured by microwave spectral studies
- It readily undergoes methoxymethylation reaction with phosphorus, carbon, oxygen and nitrogen containing nucleophiles
- It can be synthesized by reacting methylal (dimethoxymethane) with iodotrimethylsilane
Specifications
Specifications
| Molecular Formula | C9H6ClNO |
| Linear Formula | C9H6ClNO |
| MDL Number | MFCD00024011 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 179.6 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.