Learn More
Sigma Aldrich 6-Chloro-5-fluoronicotinaldehyde
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 1-bromobenzocyclobutene is a useful synthon. it has important applications in organometallic methodology. reaction between cycloheptatriene, bromoform, potassium carbonate and 18-crown-6 at 140c yields 1-bromobenzocyclobutene.1-bromobenzocyclobutene may be used in the synthesis of following compounds:
- Five-membered zirconacycles
- Benzocyclobutenol and benzocyclobutenone
- 2,3-dimethoxyprotoberberinium bromide, via reaction with 3,4-dihydro-6,7-dimethoxyisoquinoline
Specifications
Specifications
| Molecular Formula | C6H3ClFNO |
| Linear Formula | C6H3ClFNO |
| MDL Number | MFCD13189060 |
| Quantity | 1 g |
| Molecular Weight (g/mol) | 159.55 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.